(R)-2-(1-Pyrrolidinyl)butyric acid
Lieferant: Apollo Scientific
OR471424-1G
APOSOR471424-1GEA
804
EUR
APOSOR471424-1G
(R)-2-(1-Pyrrolidinyl)butyric acid
(R)-2-(1-Pyrrolidinyl)butyric acid
Formel:
C₈H₁₅NO₂ Molecular Weight: 157,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977411 CAS-Nummer: 1932770-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...