(S)-3-Phenyl-2-(1-pyrrolidinyl)propanoic acid

Lieferant: Apollo Scientific

OR471426-1G
APOSOR471426-1GEA 804 EUR
APOSOR471426-1G
(S)-3-Phenyl-2-(1-pyrrolidinyl)propanoic acid
(S)-3-Phenyl-2-(1-pyrrolidinyl)propanoic acid
Formel: C₁₃H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD24491851
CAS-Nummer: 154179-97-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR