4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Lieferant: Apollo Scientific
OR471428-1G
APOSOR471428-1GEA
804
EUR
APOSOR471428-1G
4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Formel:
C₁₀H₁₉NO₂ Molecular Weight: 185,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09035339 CAS-Nummer: 937669-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...