3-Methyl-2-(1-pyrrolidinyl)butyric acid

Lieferant: Apollo Scientific

OR471429-1G
APOSOR471429-1GEA 804 EUR
APOSOR471429-1G
3-Methyl-2-(1-pyrrolidinyl)butyric acid
3-Methyl-2-(1-pyrrolidinyl)butyric acid
Formel: C₉H₁₇NO₂
Molecular Weight: 171,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09035340
CAS-Nummer: 937652-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR