2-(1-Pyrrolidinyl)butyric acid

Lieferant: Apollo Scientific

OR471430-1G
APOSOR471430-1GEA 804 EUR
APOSOR471430-1G
2-(1-Pyrrolidinyl)butyric acid
2-(1-Pyrrolidinyl)butyric acid
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09971283
CAS-Nummer: 857206-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR