4-Methyl-1,3-oxazolidin-2-one

Lieferant: Apollo Scientific

OR471432-1G
APOSOR471432-1GEA 489 EUR
APOSOR471432-1G
4-Methyl-1,3-oxazolidin-2-one
4-Methyl-1,3-oxazolidin-2-one
Formel: C₄H₇NO₂
Molecular Weight: 101,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20502409
CAS-Nummer: 16112-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR