2-(Cyclopropylamino)-3-hydroxy-3-methylbutyric acid

Lieferant: Apollo Scientific

OR471433-1G
APOSOR471433-1GEA 1010 EUR
APOSOR471433-1G
2-(Cyclopropylamino)-3-hydroxy-3-methylbutyric acid
2-(Cyclopropylamino)-3-hydroxy-3-methylbutyric acid
Formel: C₈H₁₅NO₃
Molecular Weight: 173,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977406
CAS-Nummer: 2006276-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR