3-Hydroxy-2-(isopropylamino)-3-methylbutyric acid
Lieferant: Apollo Scientific
OR471434-1G
APOSOR471434-1GEA
1010
EUR
APOSOR471434-1G
3-Hydroxy-2-(isopropylamino)-3-methylbutyric acid
3-Hydroxy-2-(isopropylamino)-3-methylbutyric acid
Formel:
C₈H₁₇NO₃ Molecular Weight: 175,225 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977405 CAS-Nummer: 2006276-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...