1-Acetyl-3-(isopropylamino)piperidine

Lieferant: Apollo Scientific

OR471437-1G
APOSOR471437-1GEA 1540 EUR
APOSOR471437-1G
1-Acetyl-3-(isopropylamino)piperidine
1-Acetyl-3-(isopropylamino)piperidine
Formel: C₁₀H₂₀N₂O
Molecular Weight: 184,279 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098064
CAS-Nummer: 1353973-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR