(R)-2-(4-Chlorophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

OR471441-1G
APOSOR471441-1GEA 804 EUR
APOSOR471441-1G
(R)-2-(4-Chlorophenyl)-2-hydroxypropionic acid
(R)-2-(4-Chlorophenyl)-2-hydroxypropionic acid
Formel: C₉H₉ClO₃
Molecular Weight: 200,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663599
CAS-Nummer: 887304-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR