(R)-2-(2-Chlorophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

OR471443-1G
APOSOR471443-1GEA 804 EUR
APOSOR471443-1G
(R)-2-(2-Chlorophenyl)-2-hydroxypropionic acid
(R)-2-(2-Chlorophenyl)-2-hydroxypropionic acid
Formel: C₉H₉ClO₃
Molecular Weight: 200,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05663617
CAS-Nummer: 1215178-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR