2-(2-Chlorophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

OR471444-1G
APOSOR471444-1GEA 804 EUR
APOSOR471444-1G
2-(2-Chlorophenyl)-2-hydroxypropionic acid
2-(2-Chlorophenyl)-2-hydroxypropionic acid
Formel: C₉H₉ClO₃
Molecular Weight: 200,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111678
CAS-Nummer: 171202-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR