2-(4-Chlorophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

OR471445-1G
APOSOR471445-1GEA 734 EUR
APOSOR471445-1G
2-(4-Chlorophenyl)-2-hydroxypropionic acid
2-(4-Chlorophenyl)-2-hydroxypropionic acid
Formel: C₉H₉ClO₃
Molecular Weight: 200,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052509
CAS-Nummer: 4445-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR