(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

OR471448-5G
APOSOR471448-5GEA 1010 EUR
APOSOR471448-5G
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
(R)-3-(3-Bromophenyl)-2-hydroxypropionic acid
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977373
CAS-Nummer: 1932233-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR