3-Chloro-7H-pyrrolo[2,3-c]pyridazine

Lieferant: Apollo Scientific

OR471450-1G
APOSOR471450-1GEA 1220 EUR
APOSOR471450-1G
3-Chloro-7H-pyrrolo[2,3-c]pyridazine
3-Chloro-7H-pyrrolo[2,3-c]pyridazine
Formel: C₆H₄ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD17169850
CAS-Nummer: 1207625-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR