4,6-Dichloro-1H-imidazo[4,5-c]pyridin-2(3H)-one
Lieferant: Apollo Scientific
OR471471-250MG
APOSOR471471-250MGEA
31.9
EUR
APOSOR471471-250MG
4,6-Dichloro-1H-imidazo[4,5-c]pyridin-2(3H)-one
4,6-Dichloro-1H-imidazo[4,5-c]pyridin-2(3H)-one
Formel:
C₆H₃Cl₂N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD20528104 CAS-Nummer: 668268-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...