4-Chloro-2-hydroxy-1H-imidazo[4,5-c]pyridine

Lieferant: Apollo Scientific

OR471478-250MG
APOSOR471478-250MGEA 916 EUR
APOSOR471478-250MG
4-Chloro-2-hydroxy-1H-imidazo[4,5-c]pyridine
4-Chloro-2-hydroxy-1H-imidazo[4,5-c]pyridine
Formel: C₆H₄ClN₃O
Molecular Weight: 169.568 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28645819
CAS-Nummer: 54221-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR