4-Guanidino-2-methoxybenzoic acid hydrochloride

Lieferant: Apollo Scientific

OR471480-1G
APOSOR471480-1GEA 1630 EUR
APOSOR471480-1G
4-Guanidino-2-methoxybenzoic acid hydrochloride
4-Guanidino-2-methoxybenzoic acid hydrochloride
Formel: C₉H₁₁N₃O₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD29035074
CAS-Nummer: 1028486-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR