Tetrakis(4-bromophenyl)methane

Lieferant: Apollo Scientific

OR471493-5G
APOSOR471493-5GEA 226 EUR
APOSOR471493-5G
Tetrakis(4-bromophenyl)methane
Tetrakis(4-bromophenyl)methane
Formel: C₂₅H₁₆Br₄
Molecular Weight: 636.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19688472
CAS-Nummer: 105309-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR