4-Chloro-5-methyl-5H-pyrrolo[2,3-d]pyrimidin-6(7H)-one

Lieferant: Apollo Scientific
Achtung

OR471494-1G
APOSOR471494-1GEA 1390 EUR
APOSOR471494-1G
4-Chloro-5-methyl-5H-pyrrolo[2,3-d]pyrimidin-6(7H)-one
4-Chloro-5-methyl-5H-pyrrolo[2,3-d]pyrimidin-6(7H)-one
Formel: C₇H₆ClN₃O
Molecular Weight: 183,595 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24615586
CAS-Nummer: 1226804-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR