[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
Lieferant: Apollo Scientific
OR471509-1G
OR471509-250MG
APOSOR471509-1GEA
916
EUR
APOSOR471509-1G
APOSOR471509-250MG
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
[6-(3,5-Dichlorophenyl)-3-pyridyl]methanol
Formel:
C₁₂H₉Cl₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06410460 CAS-Nummer: 887974-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...