2-(4-Bromophenyl)oxazolo[4,5-c]pyridine

Lieferant: Apollo Scientific

OR471513-1G OR471513-250MG OR471513-5G
APOSOR471513-1GEA 499 EUR
APOSOR471513-1G APOSOR471513-250MG APOSOR471513-5G
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
2-(4-Bromophenyl)oxazolo[4,5-c]pyridine
Formel: C₁₂H₇BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD22573880
CAS-Nummer: 934331-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR