5-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)furan-2-carbaldehyde

Lieferant: Apollo Scientific

OR471522-5G
APOSOR471522-5GEA 1210 EUR
APOSOR471522-5G
5-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)furan-2-carbaldehyde
5-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)furan-2-carbaldehyde
Formel: C₁₀H₁₀BNO₆
Molecular Weight: 251,001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546038
CAS-Nummer: 1104637-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR