2-(6-Bromo-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Lieferant: Apollo Scientific

OR471524-5G
APOSOR471524-5GEA 1210 EUR
APOSOR471524-5G
2-(6-Bromo-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
2-(6-Bromo-3-pyridyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Formel: C₁₀H₁₀BBrN₂O₄
Molecular Weight: 312,912 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11215211
CAS-Nummer: 1816951-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR