5-Chloro-2,4-dimethoxypyrimidine

Lieferant: Apollo Scientific

OR471528-5G OR471528-1G OR471528-250MG
APOSOR471528-5GEA 364 EUR
APOSOR471528-5G APOSOR471528-1G APOSOR471528-250MG
5-Chloro-2,4-dimethoxypyrimidine
5-Chloro-2,4-dimethoxypyrimidine
Formel: C₆H₇ClN₂O₂
Molecular Weight: 174.585 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13619877
CAS-Nummer: 123551-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR