2-(3-Aminopropoxy)benzonitril

Lieferant: Apollo Scientific

OR471529-1G OR471529-5G
APOSOR471529-1GEA 256 EUR
APOSOR471529-1G APOSOR471529-5G
2-(3-Aminopropoxy)benzonitril
2-(3-Aminopropoxy)benzonitril
Formel: C₁₀H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD16495892
CAS-Nummer: 444574-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR