N1-Boc-5-methoxy-1,2-benzenediamine
Lieferant: Apollo Scientific
|
Achtung
|
OR471532-1G
OR471532-250MG
APOSOR471532-1GEA
284
EUR
APOSOR471532-1G
APOSOR471532-250MG
N1-Boc-5-methoxy-1,2-benzenediamine
N1-Boc-5-methoxy-1,2-benzenediamine
Formel:
C₁₂H₁₈N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09701255 CAS-Nummer: 362670-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...