Ethyl 3-(Boc-amino)pyrrolidine-3-carboxylate

Lieferant: Apollo Scientific

OR471535-1G OR471535-250MG
APOSOR471535-1GEA 1330 EUR
APOSOR471535-1G APOSOR471535-250MG
Ethyl 3-(Boc-amino)pyrrolidine-3-carboxylate
Formel: C₁₂H₂₂N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD29055297
CAS-Nummer: 1613023-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR