β-[(5-Bromo-3-pyridyl)oxy]phenethylamine

Lieferant: Apollo Scientific

OR471540-250MG
APOSOR471540-250MGEA 713 EUR
APOSOR471540-250MG
β-[(5-Bromo-3-pyridyl)oxy]phenethylamine
β-[(5-Bromo-3-pyridyl)oxy]phenethylamine
Formel: C₁₃H₁₃BrN₂O
Molecular Weight: 293.159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19098106
CAS-Nummer: 1468856-36-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR