4-(Benzyloxy)-3-bromophenylacetic acid

Lieferant: Apollo Scientific

Synonyme: 2-(4-(Benzyloxy)-3-bromophenyl)acetic acid

OR471558-1G
APOSOR471558-1GEA 75.4 EUR
APOSOR471558-1G
4-(Benzyloxy)-3-bromophenylacetic acid
4-(Benzyloxy)-3-bromophenylacetic acid
Formel: C₁₅H₁₃BrO₃
Molecular Weight: 321,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664748
CAS-Nummer: 5884-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR