4-Butoxy-3-hydroxyphenylacetic acid

Lieferant: Apollo Scientific

OR471561-1G
APOSOR471561-1GEA 1010 EUR
APOSOR471561-1G
4-Butoxy-3-hydroxyphenylacetic acid
4-Butoxy-3-hydroxyphenylacetic acid
Formel: C₁₂H₁₆O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD29065904
CAS-Nummer: 2006277-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR