4-(Benzyloxy)-3-hydroxyphenylacetic acid

Lieferant: Apollo Scientific

OR471563-1G
APOSOR471563-1GEA 1010 EUR
APOSOR471563-1G
4-(Benzyloxy)-3-hydroxyphenylacetic acid
4-(Benzyloxy)-3-hydroxyphenylacetic acid
Formel: C₁₅H₁₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16293773
CAS-Nummer: 28988-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR