4-Ethoxy-3-hydroxyphenylacetic acid
Lieferant: Apollo Scientific
OR471565-1G
APOSOR471565-1GEA
1010
EUR
APOSOR471565-1G
4-Ethoxy-3-hydroxyphenylacetic acid
4-Ethoxy-3-hydroxyphenylacetic acid
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016830 CAS-Nummer: 26691-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...