(R)-1-Boc-5-(bromomethyl)pyrrolidin-2-one
Lieferant: Apollo Scientific
OR471566-1G
APOSOR471566-1GEA
1210
EUR
APOSOR471566-1G
(R)-1-Boc-5-(bromomethyl)pyrrolidin-2-one
(R)-1-Boc-5-(bromomethyl)pyrrolidin-2-one
Formel:
C₁₀H₁₆BrNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28396438 CAS-Nummer: 128811-38-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...