2-Chloro-4-cyanophenylboronic acid pinacol ester

Lieferant: Apollo Scientific

OR471571-1G OR471571-5G
APOSOR471571-1GEA 127 EUR
APOSOR471571-1G APOSOR471571-5G
2-Chloro-4-cyanophenylboronic acid pinacol ester
2-Chloro-4-cyanophenylboronic acid pinacol ester
Formel: C₁₃H₁₅BClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18072641
CAS-Nummer: 945391-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR