2-Chloro-3-iodopyrazine

Lieferant: Apollo Scientific

OR471572-1G OR471572-5G
APOSOR471572-1GEA 177 EUR
APOSOR471572-1G APOSOR471572-5G
2-Chloro-3-iodopyrazine
2-Chloro-3-iodopyrazine
Formel: C₄H₂ClIN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09909706
CAS-Nummer: 191340-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR