2,3-Diamino-6-chloro-benzonitrile

Lieferant: Apollo Scientific

Synonyme: 2,3-Diamino-6-chlorobenzonitrile

OR471585-1G
APOSOR471585-1GEA 0 EUR
APOSOR471585-1G
2,3-Diamino-6-chloro-benzonitrile
2,3-Diamino-6-chloro-benzonitrile
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20723943
CAS-Nummer: 548457-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR