4-(1,3-Dioxolan-2-yl)benzeneboronic acid

Lieferant: Apollo Scientific

OR471586-250MG
APOSOR471586-250MGEA 397 EUR
APOSOR471586-250MG
4-(1,3-Dioxolan-2-yl)benzeneboronic acid
4-(1,3-Dioxolan-2-yl)benzeneboronic acid
Formel: C₉H₁₁BO₄
Molecular Weight: 193,992 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26401984
CAS-Nummer: 1401222-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR