Methyl-5-methoxy-2-methyl-1,3-benzoxazole-7-carboxylate

Lieferant: Apollo Scientific

OR471588-1G
APOSOR471588-1GEA 538 EUR
APOSOR471588-1G
Methyl-5-methoxy-2-methyl-1,3-benzoxazole-7-carboxylate
Methyl-5-methoxy-2-methyl-1,3-benzoxazole-7-carboxylate
Formel: C₁₁H₁₁NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16140362
CAS-Nummer: 1221792-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR