Methyl-7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate

Lieferant: Apollo Scientific

OR471589-1G
APOSOR471589-1GEA 521 EUR
APOSOR471589-1G
Methyl-7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate
Methyl-7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate
Formel: C₁₁H₁₁NO₄
Molecular Weight: 221,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14584796
CAS-Nummer: 1197944-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR