2-Brom-1,3,5-triisopropylbenzol 97%

Lieferant: Apollo Scientific

Synonyme: 1-Brom-2,4,6-triisopropylbenzol

OR471599-100G
APOSOR471599-100GEA 105 EUR
APOSOR471599-100G
2-Brom-1,3,5-triisopropylbenzol 97%
2-Brom-1,3,5-triisopropylbenzol
Formel: C₁₅H₂₃Br
Molecular Weight: 283,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051547
CAS-Nummer: 21524-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR