D-Valin-methylester-hydrochlorid 98%

Lieferant: Apollo Scientific

Synonyme: H-D-Val-OMe.HCl, H-D-Val-OMe·HCl

OR471600-100G
APOSOR471600-100GEA 60.9 EUR
APOSOR471600-100G
D-Valin-methylester-hydrochlorid 98%
D-Valine methyl ester hydrochloride
Formel: C₆H₁₃NO₂
Molecular Weight: 167,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00237309
CAS-Nummer: 7146-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR