β-Alaninamide hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
3-Aminopropionamide Hydrochloride, H-β-Ala-NH₂·HCl, H-β-Ala-NH2.HCl, H-β-Ala-NH2·HCl
OR471602-100G
APOSOR471602-100GEA
235
EUR
APOSOR471602-100G
β-Alaninamide hydrochloride 95%
beta-Alaninamide hydrochloride
Formel:
C₃H₈N₂O·HCl Molecular Weight: 124,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060747 CAS-Nummer: 64017-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...