2,3-Dichlormaleinsäureanhydrid 97%

Lieferant: Apollo Scientific

OR471604-100G
APOSOR471604-100GEA 289 EUR
APOSOR471604-100G
2,3-Dichlormaleinsäureanhydrid 97%
2,3-Dichlormaleinsäureanhydrid
Formel: C₄Cl₂O₃
Molecular Weight: 166,9500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005520
CAS-Nummer: 1122-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR