Boc-glycinamide 95%

Lieferant: Apollo Scientific

Synonyme: Boc-Gly-NH2, tert-Butyl (2-amino-2-oxoethyl)carbamate, N-Boc-glycinamide

OR471605-100G
APOSOR471605-100GEA 81.2 EUR
APOSOR471605-100G
Boc-glycinamide 95%
Boc-glycinamide
Formel: C₇H₁₄N₂O₃
Molecular Weight: 174,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275106
CAS-Nummer: 35150-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR