Fmoc-D-Phg-OH 98%
Lieferant: Apollo Scientific
Synonyme:
Fmoc-D-Phg-OH, N-Fmoc-D-α-phenylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-2-phenylglycine, N-Fmoc-D-2-phenylglycine
OR471607-100G
APOSOR471607-100GEA
179
EUR
APOSOR471607-100G
Fmoc-D-Phg-OH 98%
Fmoc-D-Phg-OH
Formel:
C₂₃H₁₉NO₄ Molecular Weight: 373,408 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190891 CAS-Nummer: 111524-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...