4-Methyl-2-phenylimidazole 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methyl-2-phenyl-1H-imidazole

OR471612-100G
APOSOR471612-100GEA 68.3 EUR
APOSOR471612-100G
4-Methyl-2-phenylimidazole 95%
4-Methyl-2-phenylimidazole
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047015
CAS-Nummer: 827-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR