Ethyl-7-chlor-2-oxoheptanoat 97%
Lieferant: Apollo Scientific
Synonyme:
7-Chloro-2-oxoheptanoic Acid Ethyl Ester, Ethyl 7-chloro-2-oxoheptanoate
OR471613-100G
APOSOR471613-100GEA
82.8
EUR
APOSOR471613-100G
Ethyl-7-chlor-2-oxoheptanoat 97%
Ethyl-7-chloro-2-oxoheptanoate
Formel:
C₉H₁₅ClO₃ Molecular Weight: 206.67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07698675 CAS-Nummer: 78834-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...