2-Bromo-7-iodofluorene 98%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-7-iodo-9H-fluorene

OR471614-100G
APOSOR471614-100GEA 95.7 EUR
APOSOR471614-100G
2-Bromo-7-iodofluorene 98%
2-Bromo-7-iodofluorene
Formel: C₁₃H₈BrI
Molecular Weight: 371.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024276
CAS-Nummer: 123348-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR