(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
(H-Cystine-OMe)₂·2HCl, L-Cystine dimethyl ester dihydrochloride, (H-Cys-OMe)₂·2 HCl, (H-Cys-OMe)2.2HCl
OR471615-100G
APOSOR471615-100GEA
120
EUR
APOSOR471615-100G
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 95%
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride
Formel:
C₈H₁₈Cl₂N₂O₄S₂ Molecular Weight: 341,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012490 CAS-Nummer: 32854-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...